Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610893 MIM: 601742 MIM: 603173 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHMP2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHMP2A - charged multivesicular body protein 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6807 - microRNA 6807 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM28 - tripartite motif containing 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005762.2 | Intron | NP_005753.1 |
UBE2M - ubiquitin conjugating enzyme E2 M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB45 - zinc finger and BTB domain containing 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |