Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAACAGTAGACAGTCGGTAGGT[A/G]AATCTCCCAGATCCAGAGCTACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603781 MIM: 610218 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RECQL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RECQL5 - RecQ like helicase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAP30BP - SAP30 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301839.1 | Intron | NP_001288768.1 | ||||
NM_001301855.1 | Intron | NP_001288784.1 | ||||
NM_013260.7 | Intron | NP_037392.1 | ||||
XM_011524693.2 | Intron | XP_011522995.1 | ||||
XM_011524694.2 | Intron | XP_011522996.1 | ||||
XM_011524695.2 | Intron | XP_011522997.1 | ||||
XM_017024545.1 | Intron | XP_016880034.1 | ||||
XM_017024546.1 | Intron | XP_016880035.1 | ||||
XM_017024547.1 | Intron | XP_016880036.1 | ||||
XM_017024548.1 | Intron | XP_016880037.1 | ||||
XM_017024549.1 | Intron | XP_016880038.1 | ||||
XM_017024550.1 | Intron | XP_016880039.1 |