Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATGAGTTCAGAGAACAGATTA[A/G]CTTTGGTTGCCTTGTTGGAGAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612454 MIM: 605570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEGF11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEGF11 - multiple EGF like domains 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032445.2 | 4416 | Intron | NP_115821.2 | |||
XM_017022670.1 | 4416 | Intron | XP_016878159.1 | |||
XM_017022671.1 | 4416 | UTR 3 | XP_016878160.1 | |||
XM_017022672.1 | 4416 | Intron | XP_016878161.1 | |||
XM_017022673.1 | 4416 | Intron | XP_016878162.1 | |||
XM_017022674.1 | 4416 | Intron | XP_016878163.1 | |||
XM_017022675.1 | 4416 | Intron | XP_016878164.1 |
RAB11A - RAB11A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |