Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTTCAAATTGGTAAGTGCAATG[A/T]AGAGCTCCTAACACTGGAAGTGGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605994 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP39A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP39A1 - cytochrome P450 family 39 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278738.1 | Intron | NP_001265667.1 | ||||
NM_001278739.1 | Intron | NP_001265668.1 | ||||
NM_016593.4 | Intron | NP_057677.2 | ||||
XM_005249171.3 | Intron | XP_005249228.1 | ||||
XM_011514658.1 | Intron | XP_011512960.1 | ||||
XM_017010921.1 | Intron | XP_016866410.1 | ||||
XM_017010922.1 | Intron | XP_016866411.1 | ||||
XM_017010923.1 | Intron | XP_016866412.1 | ||||
XM_017010924.1 | Intron | XP_016866413.1 | ||||
XM_017010925.1 | Intron | XP_016866414.1 |
Set Membership: |
HapMap |