Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTCTTGTCGCGGCCGCGGCGAG[A/T]TAACATCGTTTTTCCAATCTGTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601402 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100996447 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100996447 - uncharacterized LOC100996447 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MLF1 - myeloid leukemia factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130156.2 | 83 | UTR 5 | NP_001123628.1 | |||
NM_001130157.2 | 83 | UTR 5 | NP_001123629.1 | |||
NM_001195432.1 | 83 | UTR 5 | NP_001182361.1 | |||
NM_001195433.1 | 83 | UTR 5 | NP_001182362.1 | |||
NM_001195434.1 | 83 | UTR 5 | NP_001182363.1 | |||
NM_022443.4 | 83 | UTR 5 | NP_071888.1 | |||
XM_005247479.4 | 83 | UTR 5 | XP_005247536.1 | |||
XM_005247480.2 | 83 | UTR 5 | XP_005247537.1 | |||
XM_006713645.2 | 83 | UTR 5 | XP_006713708.1 | |||
XM_011512852.2 | 83 | UTR 5 | XP_011511154.1 | |||
XM_011512853.2 | 83 | UTR 5 | XP_011511155.1 | |||
XM_011512854.2 | 83 | UTR 5 | XP_011511156.1 |
Set Membership: |
HapMap |