Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTGTCGCGGCCGCGGCGAGTTA[A/G]CATCGTTTTTCCAATCTGTCCGCGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601402 | |||||||||||||||||||||||
Literature Links: |
LOC100996447 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LOC100996447 - uncharacterized LOC100996447 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MLF1 - myeloid leukemia factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130156.2 | 86 | UTR 5 | NP_001123628.1 | |||
NM_001130157.2 | 86 | UTR 5 | NP_001123629.1 | |||
NM_001195432.1 | 86 | UTR 5 | NP_001182361.1 | |||
NM_001195433.1 | 86 | UTR 5 | NP_001182362.1 | |||
NM_001195434.1 | 86 | UTR 5 | NP_001182363.1 | |||
NM_022443.4 | 86 | UTR 5 | NP_071888.1 | |||
XM_005247479.4 | 86 | UTR 5 | XP_005247536.1 | |||
XM_005247480.2 | 86 | UTR 5 | XP_005247537.1 | |||
XM_006713645.2 | 86 | UTR 5 | XP_006713708.1 | |||
XM_011512852.2 | 86 | UTR 5 | XP_011511154.1 | |||
XM_011512853.2 | 86 | UTR 5 | XP_011511155.1 | |||
XM_011512854.2 | 86 | UTR 5 | XP_011511156.1 |