Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGGGGCTCCCCTACTCACTGT[C/G]CCGTTGATGCCACCTGGCTGCGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300002 MIM: 300180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARSD - arylsulfatase D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ARSE - arylsulfatase E (chondrodysplasia punctata 1) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000047.2 | 1428 | Intron | NP_000038.2 | |||
NM_001282628.1 | 1428 | Intron | NP_001269557.1 | |||
NM_001282631.1 | 1428 | Missense Mutation | CAC,GAC | H,D 425 | NP_001269560.1 | |
XM_005274518.2 | 1428 | Intron | XP_005274575.1 | |||
XM_005274519.4 | 1428 | Intron | XP_005274576.1 | |||
XM_005274521.4 | 1428 | Intron | XP_005274578.1 | |||
XM_011545521.1 | 1428 | Intron | XP_011543823.1 | |||
XM_017029525.1 | 1428 | Intron | XP_016885014.1 | |||
XM_017029526.1 | 1428 | Intron | XP_016885015.1 |