Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTAAAGTGGAATTGCATTGGGTC[A/G]TCTGTAGACAGTGAGATCATTAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMPD1 - adenosine monophosphate deaminase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000036.2 | 2094 | Silent Mutation | GAC,GAT | D,D 682 | NP_000027.2 | |
NM_001172626.1 | 2094 | Silent Mutation | GAC,GAT | D,D 678 | NP_001166097.1 |
DENND2C - DENN domain containing 2C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |