Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCCAAAGAAGTCTCTCAGGGAA[A/G]GCCAATAAATCAGATCAGGGACAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607629 MIM: 604832 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APH1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APH1A - aph-1 homolog A, gamma-secretase subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077628.2 | Intron | NP_001071096.1 | ||||
NM_001243771.1 | Intron | NP_001230700.1 | ||||
NM_001243772.1 | Intron | NP_001230701.1 | ||||
NM_016022.3 | Intron | NP_057106.2 | ||||
XM_017001417.1 | Intron | XP_016856906.1 |
C1orf54 - chromosome 1 open reading frame 54 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CA14 - carbonic anhydrase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |