Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGTGAATACAGAGGGGCATCT[C/T]GTGTCTTGCTTGTCTGTCCTACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602201 MIM: 616092 MIM: 612805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ECM1 - extracellular matrix protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202858.1 | Intron | NP_001189787.1 | ||||
NM_004425.3 | Intron | NP_004416.2 | ||||
NM_022664.2 | Intron | NP_073155.2 |
FALEC - focally amplified long non-coding RNA in epithelial cancer | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TARS2 - threonyl-tRNA synthetase 2, mitochondrial (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |