Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCGCAGGTCATGGCAGCGCCAGG[A/C]GGCAGGTCGGAGCCGCCGCAGCTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605646 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC26A4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC26A4 - solute carrier family 26 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000441.1 | 18 | Silent Mutation | GGA,GGC | G,G 5 | NP_000432.1 | |
XM_005250425.2 | 18 | Silent Mutation | GGA,GGC | G,G 5 | XP_005250482.1 | |
XM_006716025.3 | 18 | Silent Mutation | GGA,GGC | G,G 5 | XP_006716088.1 | |
XM_017012318.1 | 18 | Silent Mutation | GGA,GGC | G,G 5 | XP_016867807.1 |
SLC26A4-AS1 - SLC26A4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |