Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAAGGGAAAGGGGATGGGGTGGC[C/T]CTTGTGGTAAGGTTAGGGGAGTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614716 MIM: 605688 MIM: 162080 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARMIL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARMIL3 - capping protein regulator and myosin 1 linker 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CPNE6 - copine 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280558.1 | Intron | NP_001267487.1 | ||||
NM_006032.3 | Intron | NP_006023.1 | ||||
XM_005268216.1 | Intron | XP_005268273.1 | ||||
XM_005268217.1 | Intron | XP_005268274.1 |
NRL - neural retina leucine zipper | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |