Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGAAGTGGCAATTGAGTGGGGGG[A/T]TGGATAAGTTCCTGACTCTGTTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300332 MIM: 300084 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGB1BP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGB1BP2 - integrin subunit beta 1 binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303277.1 | Intron | NP_001290206.1 | ||||
NM_012278.2 | Intron | NP_036410.1 | ||||
XM_017029405.1 | Intron | XP_016884894.1 |
NONO - non-POU domain containing, octamer-binding | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |