Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGTGGGGAACAAGGTGGCACAGG[A/C]CGGGGGAGAGGGAGCACAGGCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607309 MIM: 600927 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1M2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP1M2 - adaptor related protein complex 1 mu 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300887.1 | Intron | NP_001287816.1 | ||||
NM_005498.4 | Intron | NP_005489.2 |
CDKN2D - cyclin dependent kinase inhibitor 2D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRI1 - KRI1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |