Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGAGCAGGGAGACGTTTCCGGT[C/T]AAAGGGCAGAGCTCGCGTCACCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610335 MIM: 610416 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNBD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNBD2 - cyclic nucleotide binding domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207076.2 | Intron | NP_001194005.1 | ||||
NM_001304367.1 | Intron | NP_001291296.1 | ||||
NM_080834.3 | Intron | NP_543024.2 | ||||
XM_005260295.2 | Intron | XP_005260352.1 | ||||
XM_011528590.2 | Intron | XP_011526892.1 | ||||
XM_011528592.1 | Intron | XP_011526894.1 | ||||
XM_011528593.2 | Intron | XP_011526895.1 | ||||
XM_011528598.1 | Intron | XP_011526900.1 | ||||
XM_017027683.1 | Intron | XP_016883172.1 |
PHF20 - PHD finger protein 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCAND1 - SCAN domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016558.3 | Intron | NP_057642.1 | ||||
NM_033630.2 | Intron | NP_361012.2 |