Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCCTCCGGGCGCCGCTGGCCGC[C/T]TCCGTTCCCGGTCCGGGCCGCGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615956 MIM: 616743 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC151 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC151 - coiled-coil domain containing 151 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302453.1 | 2046 | UTR 3 | NP_001289382.1 | |||
NM_001302454.1 | 2046 | UTR 3 | NP_001289383.1 | |||
NM_145045.4 | 2046 | UTR 3 | NP_659482.3 | |||
XM_017026241.1 | 2046 | Intron | XP_016881730.1 |
RGL3 - ral guanine nucleotide dissociation stimulator like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |