Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAGCCCTTCGAGACCAAAAATA[C/G]CTATGATTTTCTCCACCCTCTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC14L6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC14L6 - SEC14 like lipid binding 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193336.2 | 701 | Missense Mutation | GCT,GGT | A,G 151 | NP_001180265.2 | |
XM_011530357.2 | 701 | Missense Mutation | GCT,GGT | A,G 168 | XP_011528659.1 | |
XM_011530358.2 | 701 | Intron | XP_011528660.1 | |||
XM_011530360.2 | 701 | Missense Mutation | GCT,GGT | A,G 105 | XP_011528662.1 | |
XM_017028931.1 | 701 | Missense Mutation | GCT,GGT | A,G 186 | XP_016884420.1 | |
XM_017028932.1 | 701 | Missense Mutation | GCT,GGT | A,G 97 | XP_016884421.1 | |
XM_017028933.1 | 701 | Missense Mutation | GCT,GGT | A,G 154 | XP_016884422.1 | |
XM_017028934.1 | 701 | Missense Mutation | GCT,GGT | A,G 97 | XP_016884423.1 |