Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTCCCAGCCTTTCTCTCTCTGA[A/G]GAGCATTTGCTTTGAACACCCAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607691 MIM: 611017 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAR1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SAR1A - secretion associated Ras related GTPase 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142648.1 | Intron | NP_001136120.1 | ||||
NM_020150.4 | Intron | NP_064535.1 |
TYSND1 - trypsin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |