Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAAGTGAAAATGAAGAAATTAAA[A/C]ACGAAGAAAAACCTGGAAAAAGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf73 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf73 - chromosome 2 open reading frame 73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100396.1 | 197 | Missense Mutation | AAC,CAC | N,H 29 | NP_001093866.1 | |
XM_005264133.1 | 197 | Intron | XP_005264190.1 | |||
XM_005264134.3 | 197 | Intron | XP_005264191.1 | |||
XM_011532510.2 | 197 | Missense Mutation | AAC,CAC | N,H 29 | XP_011530812.1 | |
XM_011532513.2 | 197 | Intron | XP_011530815.1 | |||
XM_017003323.1 | 197 | Intron | XP_016858812.1 | |||
XM_017003324.1 | 197 | Missense Mutation | AAC,CAC | N,H 29 | XP_016858813.1 |