Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCTGCAGCCTTGGCGGGCGGCT[C/T]GCACTCACCGCTCCTCCTGGGCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601211 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRE5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRE5 - adhesion G protein-coupled receptor E5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025160.2 | Intron | NP_001020331.1 | ||||
NM_001784.4 | Intron | NP_001775.2 | ||||
NM_078481.3 | Intron | NP_510966.1 | ||||
XM_011528451.2 | Intron | XP_011526753.1 | ||||
XM_017027547.1 | Intron | XP_016883036.1 |
DDX39A - DEAD-box helicase 39A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005804.3 | Intron | NP_005795.2 | ||||
XM_006722606.3 | Intron | XP_006722669.1 | ||||
XM_011527620.1 | Intron | XP_011525922.1 | ||||
XM_011527621.2 | Intron | XP_011525923.1 |