Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133510 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERCC3 - ERCC excision repair 3, TFIIH core complex helicase subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000122.1 | 2600 | UTR 3 | NP_000113.1 | |||
NM_001303416.1 | 2600 | UTR 3 | NP_001290345.1 | |||
NM_001303418.1 | 2600 | UTR 3 | NP_001290347.1 | |||
XM_011510794.1 | 2600 | UTR 3 | XP_011509096.1 | |||
XM_011510795.1 | 2600 | UTR 3 | XP_011509097.1 | |||
XM_017003583.1 | 2600 | UTR 3 | XP_016859072.1 |