Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCACCATTAGCTGGAGGAGAGA[C/G]GGACTGTCTGCACTTAGATATTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf70 - chromosome 16 open reading frame 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320540.1 | Intron | NP_001307469.1 | ||||
NM_001320541.1 | Intron | NP_001307470.1 | ||||
NM_001320542.1 | Intron | NP_001307471.1 | ||||
NM_001320543.1 | Intron | NP_001307472.1 | ||||
NM_025187.4 | Intron | NP_079463.2 | ||||
XM_006721284.3 | Intron | XP_006721347.1 | ||||
XM_011523350.2 | Intron | XP_011521652.1 | ||||
XM_017023729.1 | Intron | XP_016879218.1 | ||||
XM_017023730.1 | Intron | XP_016879219.1 | ||||
XM_017023731.1 | Intron | XP_016879220.1 |