Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACAAAGCTGAAGGGCTTGACGA[C/T]CTGCAGCAGCAGCTGGGAGAACTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300060 MIM: 312090 MIM: 312070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAGE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAGE3 - L antigen family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC10A3 - solute carrier family 10 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142391.2 | 1442 | Missense Mutation | ATC,GTC | I,V 325 | NP_001135863.1 | |
NM_001142392.2 | 1442 | Missense Mutation | ATC,GTC | I,V 354 | NP_001135864.1 | |
NM_019848.4 | 1442 | Missense Mutation | ATC,GTC | I,V 354 | NP_062822.1 | |
XM_005277913.4 | 1442 | Missense Mutation | ATC,GTC | I,V 409 | XP_005277970.1 | |
XM_006724847.3 | 1442 | Missense Mutation | ATC,GTC | I,V 447 | XP_006724910.1 | |
XM_006724848.3 | 1442 | Missense Mutation | ATC,GTC | I,V 418 | XP_006724911.1 | |
XM_011531200.2 | 1442 | Missense Mutation | ATC,GTC | I,V 409 | XP_011529502.2 | |
XM_011531201.2 | 1442 | Missense Mutation | ATC,GTC | I,V 354 | XP_011529503.1 |
UBL4A - ubiquitin like 4A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |