Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAACCTAGGAAATAAAAACTAGCT[A/G]CTTTTTAAGTTACACAAGATTGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612472 MIM: 614032 | ||||||||||||||||||||
Literature Links: |
METTL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
METTL3 - methyltransferase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019852.4 | 4452 | Intron | NP_062826.2 | |||
XM_006720206.3 | 4452 | Intron | XP_006720269.1 | |||
XM_011536968.1 | 4452 | Intron | XP_011535270.1 |
TOX4 - TOX high mobility group box family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303523.1 | 4452 | UTR 3 | NP_001290452.1 | |||
NM_014828.3 | 4452 | UTR 3 | NP_055643.1 |