Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTAAATCTGGAGCTGCCAAGTTA[C/G]TTAAGAGAAATTTCCTTGAGGCGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605761 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ASB4 - ankyrin repeat and SOCS box containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016116.2 | 120 | Missense Mutation | CTT,GTT | L,V 17 | NP_057200.1 | |
NM_145872.2 | 120 | Missense Mutation | CTT,GTT | L,V 17 | NP_665879.1 | |
XM_017012303.1 | 120 | Missense Mutation | CTT,GTT | L,V 17 | XP_016867792.1 |
Set Membership: |
HapMap |