Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGAAGGGCAAGTTGGGAGTAG[A/G]GGGTATGCCTGGGACACAGGTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
35 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614612 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KANK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KANK4 - KN motif and ankyrin repeat domains 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320269.1 | Intron | NP_001307198.1 | ||||
NM_181712.4 | Intron | NP_859063.3 | ||||
XM_011540843.2 | Intron | XP_011539145.1 | ||||
XM_011540845.2 | Intron | XP_011539147.1 | ||||
XM_017000483.1 | Intron | XP_016855972.1 | ||||
XM_017000484.1 | Intron | XP_016855973.1 | ||||
XM_017000485.1 | Intron | XP_016855974.1 |