Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATGGAGACGTATGCGGAGGTTGG[C/G]AAGGAGGGCAAGGTAGAGAAAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX32 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNX32 - sorting nexin 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152760.2 | 445 | Silent Mutation | GGC,GGG | G,G 8 | NP_689973.2 | |
XM_006718488.3 | 445 | Silent Mutation | GGC,GGG | G,G 8 | XP_006718551.1 | |
XM_017017470.1 | 445 | Intron | XP_016872959.1 | |||
XM_017017471.1 | 445 | UTR 5 | XP_016872960.1 | |||
XM_017017472.1 | 445 | UTR 5 | XP_016872961.1 | |||
XM_017017473.1 | 445 | UTR 5 | XP_016872962.1 |