Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCTACGTGAAGCTTGGCCTCAC[C/T]GTCGTCTACACCGTGTTCTACGCGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604658 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR137B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPR137B - G protein-coupled receptor 137B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003272.3 | 229 | Silent Mutation | ACC,ACT | T,T 46 | NP_003263.1 | |
XM_017002209.1 | 229 | Silent Mutation | ACC,ACT | T,T 46 | XP_016857698.1 | |
XM_017002210.1 | 229 | Silent Mutation | ACC,ACT | T,T 46 | XP_016857699.1 |