Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAGACAATACGATGGAACGTTCT[A/T]TGAATTTCGTACTTATTACCTTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608871 MIM: 608872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIPSNAP3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NIPSNAP3A - nipsnap homolog 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NIPSNAP3B - nipsnap homolog 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018376.3 | 610 | Missense Mutation | TAT,TTT | Y,F 37 | NP_060846.2 | |
XM_011518839.1 | 610 | Missense Mutation | TAT,TTT | Y,F 37 | XP_011517141.1 | |
XM_017014889.1 | 610 | Missense Mutation | TAT,TTT | Y,F 37 | XP_016870378.1 |