Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613814 MIM: 614535 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC19 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TTC19 - tetratricopeptide repeat domain 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271420.1 | Intron | NP_001258349.1 | ||||
NM_017775.3 | Intron | NP_060245.3 | ||||
XM_017024801.1 | Intron | XP_016880290.1 | ||||
XM_017024802.1 | Intron | XP_016880291.1 |
ZSWIM7 - zinc finger SWIM-type containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |