Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603445 MIM: 608746 MIM: 610822 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KHSRP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KHSRP - KH-type splicing regulatory protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A23 - solute carrier family 25 member 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A41 - solute carrier family 25 member 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321298.1 | 609 | Silent Mutation | CGC,CGT | R,R 105 | NP_001308227.1 | |
NM_173637.3 | 609 | Silent Mutation | CGC,CGT | R,R 243 | NP_775908.2 | |
XM_011527926.1 | 609 | Silent Mutation | CGC,CGT | R,R 179 | XP_011526228.1 |
Set Membership: |
HapMap |