Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF566 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF566 - zinc finger protein 566 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145343.1 | 1689 | UTR 3 | NP_001138815.1 | |||
NM_001145344.1 | 1689 | UTR 3 | NP_001138816.1 | |||
NM_001145345.1 | 1689 | UTR 3 | NP_001138817.1 | |||
NM_001300970.1 | 1689 | UTR 3 | NP_001287899.1 | |||
NM_032838.4 | 1689 | UTR 3 | NP_116227.1 | |||
XM_005259354.2 | 1689 | UTR 3 | XP_005259411.1 | |||
XM_005259356.2 | 1689 | UTR 3 | XP_005259413.1 | |||
XM_006723447.2 | 1689 | UTR 3 | XP_006723510.1 | |||
XM_011527428.1 | 1689 | UTR 3 | XP_011525730.1 | |||
XM_017027399.1 | 1689 | UTR 3 | XP_016882888.1 | |||
XM_017027400.1 | 1689 | UTR 3 | XP_016882889.1 |
Set Membership: |
HapMap |