Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGCAGACGCTGTGGGGAGCTCC[A/G]AAGGCCCAACAAGTGCTGCAGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf210 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf210 - chromosome 1 open reading frame 210 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164829.1 | 198 | Missense Mutation | NP_001158301.1 | |||
NM_182517.2 | 198 | Missense Mutation | NP_872323.1 | |||
XM_011540802.1 | 198 | Missense Mutation | XP_011539104.1 |
TMEM125 - transmembrane protein 125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |