Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTTCCCAATGGCCTTTCGAACC[C/T]GCAGTCGGCAAAGGTGAGCCAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605759 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB2 - ankyrin repeat and SOCS box containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202429.1 | 2088 | Missense Mutation | CAG,CGG | Q,R 603 | NP_001189358.1 | |
NM_016150.4 | 2088 | Missense Mutation | CAG,CGG | Q,R 555 | NP_057234.2 | |
XM_005267758.3 | 2088 | Missense Mutation | CAG,CGG | Q,R 603 | XP_005267815.1 | |
XM_011536834.2 | 2088 | Missense Mutation | CAG,CGG | Q,R 551 | XP_011535136.1 | |
XM_011536835.2 | 2088 | Missense Mutation | CAG,CGG | Q,R 501 | XP_011535137.1 | |
XM_017021369.1 | 2088 | Missense Mutation | CAG,CGG | Q,R 568 | XP_016876858.1 |
FAM181A - family with sequence similarity 181 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM181A-AS1 - FAM181A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |