Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAACTGCCCTGTGCCCCGTGTG[G/T]TGGGTGTGGCACCCTTACTGCTCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612793 | ||||||||||||||||||||
Literature Links: |
PTDSS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTDSS2 - phosphatidylserine synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005253153.1 | Intron | XP_005253210.1 | ||||
XM_011520391.2 | Intron | XP_011518693.1 | ||||
XM_011520392.2 | Intron | XP_011518694.1 | ||||
XM_011520393.2 | Intron | XP_011518695.1 | ||||
XM_017018368.1 | Intron | XP_016873857.1 | ||||
XM_017018369.1 | Intron | XP_016873858.1 | ||||
XM_017018370.1 | Intron | XP_016873859.1 | ||||
XM_017018371.1 | Intron | XP_016873860.1 |