Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGTCCAGCCTGGGGCCAGCTG[A/G]TAATGACATGGCCTGTTTTCTGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188390 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR1244-1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR1244-1 - microRNA 1244-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTMA - prothymosin, alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099285.1 | Intron | NP_001092755.1 | ||||
NM_002823.4 | Intron | NP_002814.3 | ||||
XM_011511518.1 | Intron | XP_011509820.1 | ||||
XM_011511519.1 | Intron | XP_011509821.1 | ||||
XM_011511520.1 | Intron | XP_011509822.1 | ||||
XM_011511521.1 | Intron | XP_011509823.1 | ||||
XM_011511522.2 | Intron | XP_011509824.1 |