Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTCTACCAGGAGGTAATCTGGG[C/T]GCACAGAATTTATCTGAGGTAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602533 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARK7 - Parkinsonism associated deglycase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123377.1 | Intron | NP_001116849.1 | ||||
NM_007262.4 | Intron | NP_009193.2 | ||||
XM_005263424.3 | Intron | XP_005263481.1 |