Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGGCCCACCCATGTCCAGCACC[C/G]TTGGAGGGCTCCTGGCCAGGAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf180 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf180 - chromosome 14 open reading frame 180 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM179 - transmembrane protein 179 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286389.1 | 631 | Intron | NP_001273318.1 | |||
NM_001286390.1 | 631 | Missense Mutation | AAC,AAG | N,K 204 | NP_001273319.1 | |
XM_011536745.2 | 631 | Intron | XP_011535047.1 | |||
XM_011536746.2 | 631 | Intron | XP_011535048.1 | |||
XM_011536750.2 | 631 | Intron | XP_011535052.1 |