Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTTAGTAGCATGAACATCTCTG[C/T]CACCATCTATCATATGAATATCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182392 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCN7A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SCN7A - sodium voltage-gated channel alpha subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002976.3 | 5394 | Missense Mutation | GAC,GGC | D,G 1657 | NP_002967.2 | |
XM_006712680.2 | 5394 | Missense Mutation | GAC,GGC | D,G 1657 | XP_006712743.1 | |
XM_006712681.3 | 5394 | Missense Mutation | GAC,GGC | D,G 1657 | XP_006712744.1 | |
XM_006712682.3 | 5394 | Missense Mutation | GAC,GGC | D,G 1657 | XP_006712745.1 | |
XM_011511615.2 | 5394 | Missense Mutation | GAC,GGC | D,G 1657 | XP_011509917.1 | |
XM_017004667.1 | 5394 | Missense Mutation | GAC,GGC | D,G 1657 | XP_016860156.1 |
Set Membership: |
HapMap |