Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTTAACCATGGCGCAGCGGGCA[A/G]TGTGGCTCATAAGCCACGAACCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614368 MIM: 604469 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP5M1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP5M1 - adaptor related protein complex 5 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018229.3 | 373 | Missense Mutation | ATG,GTG | M,V 6 | NP_060699.3 | |
XM_006720196.2 | 373 | Intron | XP_006720259.1 | |||
XM_011536940.2 | 373 | Missense Mutation | ATG,GTG | M,V 6 | XP_011535242.1 |
EXOC5 - exocyst complex component 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006544.3 | 373 | Intron | NP_006535.1 | |||
XM_005267272.3 | 373 | Intron | XP_005267329.1 |