Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGTACGGTGAGGTGGAAGAGGTA[G/A]AGATCCTCCTTCACCCCCGTACGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610930 MIM: 611052 | ||||||||||||||||||||
Literature Links: |
ORAI3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ORAI3 - ORAI calcium release-activated calcium modulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD1A - SET domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014712.2 | 461 | Missense Mutation | AAG,GAG | K,E 124 | NP_055527.1 | |
XM_005255723.1 | 461 | Missense Mutation | AAG,GAG | K,E 124 | XP_005255780.1 | |
XM_006721106.3 | 461 | Missense Mutation | AAG,GAG | K,E 124 | XP_006721169.1 | |
XM_017023909.1 | 461 | Missense Mutation | AAG,GAG | K,E 124 | XP_016879398.1 |