Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGTGGCTGCTCGCGGTCGGCGT[G/T]CCCCGACGTACAGCGGGCCGGGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611299 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
G2E3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
G2E3 - G2/M-phase specific E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308097.1 | 84 | UTR 5 | NP_001295026.1 | |||
NM_017769.4 | 84 | UTR 5 | NP_060239.2 | |||
XM_011536925.2 | 84 | Intron | XP_011535227.1 | |||
XM_011536926.1 | 84 | Intron | XP_011535228.1 |