Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAGGAGGAAAACAGGTCTGTCT[A/G]CTAGGTCAGGCATCTAGAAAATTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612060 | ||||||||||||||||||||
Literature Links: |
ZNRF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNRF1 - zinc and ring finger 1, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032268.4 | Intron | NP_115644.1 | ||||
XM_011523392.1 | Intron | XP_011521694.1 | ||||
XM_017023793.1 | Intron | XP_016879282.1 | ||||
XM_017023794.1 | Intron | XP_016879283.1 |