Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCTGGAGTTACGGACATATCGC[C/T]GCCCCTTGGGGGTTCCGGGACGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602102 MIM: 607381 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUPT5H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUPT5H - SPT5 homolog, DSIF elongation factor subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIMM50 - translocase of inner mitochondrial membrane 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011527491.2 | 59 | Missense Mutation | CCG,CTG | P,L 13 | XP_011525793.1 | |
XM_017027472.1 | 59 | Intron | XP_016882961.1 |