Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTACCCGTGTGTGTGTACGTGCA[C/T]ATACGTGTGTATGGTGGAAGCATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134636 MIM: 154950 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHURC1-FNTB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHURC1-FNTB - CHURC1-FNTB readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202558.1 | Intron | NP_001189487.1 | ||||
NM_001202559.1 | Intron | NP_001189488.1 |
FNTB - farnesyltransferase, CAAX box, beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002028.3 | Intron | NP_002019.1 |
MAX - MYC associated factor X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271068.1 | Intron | NP_001257997.1 | ||||
NM_001271069.1 | Intron | NP_001257998.1 | ||||
NM_001320415.1 | Intron | NP_001307344.1 | ||||
NM_002382.4 | Intron | NP_002373.3 | ||||
NM_145112.2 | Intron | NP_660087.1 | ||||
NM_145113.2 | Intron | NP_660088.1 | ||||
NM_145114.2 | Intron | NP_660089.1 | ||||
NM_197957.3 | Intron | NP_932061.1 | ||||
XM_011536773.2 | Intron | XP_011535075.1 | ||||
XM_017021312.1 | Intron | XP_016876801.1 | ||||
XM_017021313.1 | Intron | XP_016876802.1 |