Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTATAGGGCTCTGAGCCTGATG[A/G]AGATGGATGGGAGTGGAGTAGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609933 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
REG3G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
REG3G - regenerating family member 3 gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008387.2 | Intron | NP_001008388.1 | ||||
NM_001270040.1 | Intron | NP_001256969.1 | ||||
NM_198448.3 | Intron | NP_940850.1 | ||||
XM_005264135.2 | Intron | XP_005264192.1 |