Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGGTCTGCTGGCATATTTTTCT[A/G]AGTAGATTTATGTAGCTTAAGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182396 MIM: 600914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100289511 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100289511 - uncharacterized LOC100289511 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC10A1 - solute carrier family 10 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRSF5 - serine and arginine rich splicing factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039465.1 | Intron | NP_001034554.1 | ||||
NM_001320214.1 | Intron | NP_001307143.1 | ||||
NM_006925.4 | Intron | NP_008856.2 | ||||
XM_005267999.1 | Intron | XP_005268056.1 | ||||
XM_005268000.1 | Intron | XP_005268057.1 | ||||
XM_011537077.2 | Intron | XP_011535379.1 | ||||
XM_017021593.1 | Intron | XP_016877082.1 | ||||
XM_017021594.1 | Intron | XP_016877083.1 |