Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCTCCTCAATGTCTGTGCCCAT[A/T]TATTCCAATTTTTGATTTGGGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612763 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POGK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POGK - pogo transposable element with KRAB domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001314014.1 | Intron | NP_001300943.1 | ||||
NM_017542.4 | Intron | NP_060012.3 | ||||
XM_005245370.3 | Intron | XP_005245427.1 | ||||
XM_011509814.2 | Intron | XP_011508116.1 | ||||
XM_011509815.1 | Intron | XP_011508117.1 | ||||
XM_017001914.1 | Intron | XP_016857403.1 | ||||
XM_017001915.1 | Intron | XP_016857404.1 |
TADA1 - transcriptional adaptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |