Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACCCACCGGGCCCACACACACC[A/C]GCCTGCTGCACAAACTGCAGCACGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604670 MIM: 170998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDPF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDPF1 - cysteine rich DPF motif domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207327.4 | 633 | Missense Mutation | CGG,CTG | R,L 70 | NP_997210.3 | |
XM_011529960.1 | 633 | Missense Mutation | CGG,CTG | R,L 70 | XP_011528262.1 | |
XM_011529962.2 | 633 | Missense Mutation | CGG,CTG | R,L 70 | XP_011528264.1 | |
XM_011529963.1 | 633 | Silent Mutation | GCG,GCT | A,A 69 | XP_011528265.1 | |
XM_011529964.1 | 633 | Silent Mutation | GCG,GCT | A,A 69 | XP_011528266.1 | |
XM_011529965.2 | 633 | Missense Mutation | CGG,CTG | R,L 70 | XP_011528267.1 |
PKDREJ - polycystin (PKD) family receptor for egg jelly | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPARA - peroxisome proliferator activated receptor alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |