Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600834 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF165 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF165 - zinc finger protein 165 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003447.3 | 54 | UTR 5 | NP_003438.1 | |||
XM_017011258.1 | 54 | Intron | XP_016866747.1 | |||
XM_017011259.1 | 54 | Intron | XP_016866748.1 | |||
XM_017011260.1 | 54 | Intron | XP_016866749.1 | |||
XM_017011261.1 | 54 | Intron | XP_016866750.1 | |||
XM_017011262.1 | 54 | Intron | XP_016866751.1 |